missing translation for 'onlineSavingsMsg'
Learn More
Tetrathiafulvalene, 97%, Thermo Scientific Chemicals Product Code.: 11364096

Tetrathiafulvalene, 97%, Thermo Scientific Chemicals

Product Code. 11364096
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11364096

Brand: Thermo Scientific Alfa Aesar L19229.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.

Solubility
It is insoluble in water. Soluble in organic solvents.

Notes
Air & Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31366-25-3
Molecular Formula C6H4S4
Molecular Weight (g/mol) 204.338
MDL Number MFCD00005492
InChI Key FHCPAXDKURNIOZ-UHFFFAOYSA-N
Synonym tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithioleShow MoreShow Less
PubChem CID 99451
ChEBI CHEBI:52444
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 116°C to 120°C
Quantity 1 g
Beilstein 1617956
Sensitivity Air and light sensitive
Merck Index 14,9242
Solubility Information It is insoluble in water. Soluble in organic solvents.
Formula Weight 204.35
Percent Purity 97%
Chemical Name or Material Tetrathiafulvalene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251764 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL19229.03,L19229.MD
Lot Number10241640 Certificate TypeCertificate of Analysis Date29/06/2022 Catalog NumberL19229.03,L19229.MD
Lot NumberF13X073 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19229.03,L19229.MD
Lot NumberF24Z55 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19229.03,L19229.MD
Lot Number10153738 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL19229.03,L19229.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrathiafulvalene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.