missing translation for 'onlineSavingsMsg'
Learn More
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals Product Code.: 10479490

Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals

Product Code. 10479490
1 g, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10479490

Brand: Thermo Scientific Acros 179410010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31366-25-3
Molecular Formula C6H4S4
Molecular Weight (g/mol) 204.34
MDL Number MFCD00005492
InChI Key FHCPAXDKURNIOZ-UHFFFAOYSA-N
Synonym tetrathiafulvalene, 2-1,3-dithiol-2-ylidene-1,3-dithiole, 1,3-dithiole, 2-1,3-dithiol-2-ylidene, unii-hy1en16w9t, 1,4,5,8-tetrathiafulvalene, ttf, delta-2:2'-bis 1,3-dithiazole, hy1en16w9t, 2,2'-bi-1,3-dithiole, delta-2,2'-bi-1,3-dithioleShow MoreShow Less
PubChem CID 99451
ChEBI CHEBI:52444
IUPAC Name 2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES C1=CSC(=C2SC=CS2)S1

Specifications

Melting Point 117°C to 120°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 9390
Formula Weight 204.34
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetrathiafulvalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0399730 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number179412500,179410010
Lot NumberA0412918 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number179412500,179410010
Lot NumberA0409317 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number179412500,179410010
Lot NumberA0417644 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number179412500,179410010
Lot NumberA0447524 Certificate TypeCertificate of Analysis Date02/12/2022 Catalog Number179412500,179410010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrathiafulvalene, 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.