missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Kit

Quickly and reliably measure protein concentration of cell lysates or other samples that contain Triton™ X-100, NP-40 or other detergents using the the Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Kit.

Brand:  Thermo Scientific™ 23246

169.40 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


189.64 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 15398086

  • £224.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific Pierce Detergent Compatible Bradford Assay Kit is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation. Proprietary additives to the Bradford Reagent make it compatible with up to 1% or higher of detergents and lysis reagents that are commonly used in life science research, including Triton™ X-100 and NP-40.

  • Convenient - detergent-free standard curve
  • Flexible - compatible with samples both with and without detergent
  • Minimal sample - requires only 10μL for microplate procedure
  • Colorimetric - measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use - single reagent, no working reagent preparation required
  • Fast - 10 minute incubation at room temperature
  • Broad range - detects protein concentration in the ranges of 2 to 1500μg/mL
Similar to the Bradford method, coomassie dye binds protein in an acidic medium causing an immediate shift in absorption maximum from 465 nm to 595 nm with a concomitant color change from green to blue. In addition, the assay is complete in just 10 minutes.
The protein assay can be performed in either test tube or microplate format. The standard working range is 100-1500μg/mL with up to 1% detergent (or higher in some cases). Protein concentrations are estimated by reference to absorbances obtained for a series of standard protein dilutions, typically bovine serum albumin (BSA), which are assayed alongside the unknown samples. Because the color response with coomassie dye is non-linear with increasing protein concentration, a standard curve must be completed with each assay. Standards can be used directly without preparing them in the same detergent found in the test samples.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Detergent Compatible Bradford Assay Reagent, 450 mL, store at 4°C
Albumin Standard Ampules, 10 x 1 mL, store at room temperature
Colorimetric
Protein Quantitation Assay
Solution-based Detection, Absorbance
300 Test Tube Assays
Bradford Assay
Pierce™
Not Target-Specific
Spectrophotometer, Microplate Reader
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA399415 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number23246
Lot NumberZJ400592 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number23246
Lot NumberZI398050A Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number23246
Lot NumberZI398050 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number23246
Lot NumberZH392713 Certificate TypeCertificate of Analysis Date31/08/2024 Catalog Number23246
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Kit > Kit, 450mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.