missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peroxide Assay Reagent A

Measure total peroxide levels in either aqueous or lipid-containing laboratory reagents

Brand:  Thermo Scientific™ 23281

Product Code. 10580234

  • £80.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

These aqueous and lipid-compatible peroxide assay kits detect peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. One assay kit has an aqueous-compatible formulation that includes sorbitol, which provides sensitivity enhancement. The other assay kit has a formulation that may be used with lipid-containing samples without first extracting the lipids; however, because this kit does not contain sorbitol, the assay is less sensitive. Most proteins do not interfere with these assays, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.
In these assays, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With the aqueous-compatible formulation, peroxide first reacts with sorbitol, converting it to a peroxyl radical, which in turn initiates Fe2+ oxidation to Fe3+. In the lipid compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560nm. Peroxide levels in test samples can be determined by calculation from the know extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.

Highlights:

  • Peroxides – detects and measures hydrogen peroxide (H2O2) levels in biological and other liquids samples
  • Convenient – no proprietary reagents involved but the ready-to-use kits remove the variability, difficulty and added expense of formulating the reagents yourself
  • Versatile – quantitation by calculation from the extinction coefficient or by reference to a standard curve enables the method to be easily adapted to a variety of sample types and testing purposes
  • Two formulations – choose the aqueous-compatible kit for maximum sensitivity with non-lipid samples; choose the lipid-compatible kit for best results with lipid-containing samples

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Colorimetric
Quantitative Peroxide Assay
Solution-based Detection, Absorbance
1 mL
Peroxide Assay Reagent A
Pierce™
Not Target-Specific
Spectrophotometer, Microplate Reader
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA402848 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number23281
Lot NumberZJ401419 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number23281
Lot NumberZG396282 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number23281
Lot NumberZF391552 Certificate TypeCertificate of Analysis Date22/06/2024 Catalog Number23281
Lot NumberZD385408 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number23281
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Peroxide Assay Reagent A > Peroxoquant Reagent A 1/pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.