Learn More
Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators
Accessory Rotors
Brand:  Thermo Scientific™ RH6-25
 View more versions of this product
Product Code. 11718579
  • £1059.49 / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Easy to use circular rotors with different tube hole sizes to accommodate different sample sizes. These rotors require no additional accessories and fit directly into your concentrator.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass And Plastic Tube
Savant™ DNA SpeedVac™ concentrator rotor
6 x 30 mL Tubes
30 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Rotors for Savant™ DNA SpeedVac™ Concentrators > 6 x 30mL; 24x105mm