Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand:  SymLine™ 117111
 View more versions of this product
Product Code. 18636488
  • £112.00 / 1 sheet
Explore more special offers
Description
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
Black
Thread adapter, S60/61 (m) to S50 (f), excentric with FKM gasket
PE-HD-EL
Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SymLine™ Adapters > Description: Thread adapter, S60/61 (m) to S50 (f), excentric with FKM gasket