missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) phthalocyanine dichloride, Thermo Scientific Chemicals Product Code.: 11364537

Tin(IV) phthalocyanine dichloride, Thermo Scientific Chemicals

Product Code. 11364537
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11364537

Brand: Thermo Scientific Alfa Aesar 039606.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tin(IV) phthalocyanine dichloride is a stable metal phthalocyanine dye utilized in biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18253-54-8
Molecular Formula C32H16Cl2N8Sn
Molecular Weight (g/mol) 702.146
MDL Number MFCD00054210
InChI Key BBRNKSXHHJRNHK-UHFFFAOYSA-L
Synonym tin iv phthalocyanine dichloride, sncl2pc, dichlorotin phthalocyanine, tin phthalocyanine dichloride, tin iv phthalocyaninedichloride
PubChem CID 9580361
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C6N4[Sn](N7C(=NC2=N3)C8=CC=CC=C8C7=NC9=NC(=N6)C1=CC=CC=C19)(Cl)Cl

Specifications

Quantity 5 g
UN Number UN3146
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 702.34
Physical Form Powder
Chemical Name or Material Tin(IV) phthalocyanine dichloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ02K055 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number039606.06,039606.03
Lot NumberY02H034 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039606.06,039606.03
Lot NumberF08X073 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039606.06,039606.03
Lot NumberF28Z022 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039606.06,039606.03
Lot NumberH07R019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039606.06,039606.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) phthalocyanine dichloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.