Learn More
Topiramate, MedChemExpress
MedChemExpress Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Brand:  Medchem Express HY-B0122
Encompass_Preferred
 View more versions of this product
Product Code. 16492026
  • £356.95 / 500mg
Explore more special offers
Description
Description
Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Specifications
Specifications
Topiramate
White
C12H21NO8S
McN 4853 RWJ 17021
NS(OC[C@]1(OC(C)(C)O2)[C@@H]2[C@H](OC(C)(C)O3)[C@H]3CO1)(=O)=O
339.36
Research
Neuroscience-Neuromodulation
97240-79-4
Research
500 mg
DMSO : ≥ 100 mg/mL (294.67 mM) ∣H2O : 4 mg/mL (11.79 mM; Need ultrasonic)
339.36
98.0%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Topiramate, MedChemExpress