missing translation for 'onlineSavingsMsg'
Learn More
trans-Anethole, 99%, Thermo Scientific Chemicals Product Code.: 10149931

trans-Anethole, 99%, Thermo Scientific Chemicals

Product Code. 10149931
5 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10149931

Brand: Thermo Scientific Acros 152530050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4180-23-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009284
InChI Key RUVINXPYWBROJD-ONEGZZNKSA-N
Synonym anethole, trans-anethole, e-anethole, 4-propenylanisole, anise camphor, trans-anethol, isoestragole, e-1-methoxy-4-prop-1-en-1-yl benzene, anethol, p-anetholeShow MoreShow Less
PubChem CID 637563
ChEBI CHEBI:35616
IUPAC Name 1-methoxy-4-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=C(C=C1)OC

Specifications

Melting Point 23.0°C
Color Colorless to Yellow
Density 0.9875g/mL
Boiling Point 234.0°C to 237.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHC6H4OCH3
Refractive Index 1.5590 to 1.5620
Quantity 5 mL
Beilstein 06, 566
Merck Index 15, 636
Specific Gravity 0.9875
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with ether, chloroform, soluble in benzene, ethyl acetate, acetone,, alcohol and carbon disulfide
Formula Weight 148.2
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material trans-Anethole, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463476 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number152535000,152530050,152531000
Lot NumberA0463476 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number152535000,152530050,152531000
Lot NumberA0456057 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number152535000,152530050,152531000
Lot NumberA0450229 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number152535000,152530050,152531000
Lot NumberA0459922 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number152535000,152530050,152531000
Product Identifier
  • trans-Anethole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-Anethole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.