Learn More
BRAND™ Single-Channel Micropipette Transferpette™ S, Fixed-Volume, DE-M

Efficient: one-hand operation. Flexible: ergonomic design for all hand sizes.
Brand: BRAND™ 705828
121.29 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
The Transferpette™ S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette™ S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.
- The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment.
- The Transferpette™ S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools.
- A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
- Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.

Specifications
DIN EN 285 | |
Autoclavable | |
Fixed Volume Pipette | |
15 to 40°C | |
11.3 in. | |
4.7 in. | |
1.9 in. |
Transferpette™ S, 50 μL, fixed volume, F-50, DE-M | |
Non-sterile | |
50 ÎĽL | |
36 Months | |
28.7 cm | |
12 cm | |
4.8 cm |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Brand Transferpette S (EN)
Accessories and Services
-
BRAND™ Pipette Tips, 2 - 200 μL, PP
X480 PIPETTE TIP TIPBOX 2-200UL PP Product Code: 15104917ÂŁ45.10 / Pack of 480 -
BRAND™ Pipette Tips, 2 - 200 μL, PP X1000 PIPETTE TIP BULK 2-200UL PP Product Code: 15154887
ÂŁ41.20 / Pack of 1000
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
17Hours
15Minutes
15Seconds
54New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.