missing translation for 'onlineSavingsMsg'
Learn More
Triethylene glycol, 99%, Thermo Scientific Chemicals Product Code.: 11409682

Triethylene glycol, 99%, Thermo Scientific Chemicals

Product Code. 11409682
2500 mL
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy

Product Code. 11409682

Brand: Thermo Scientific Alfa Aesar A10772.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

Solubility
Miscible with water, ethanol, acetone, benzene, toluene, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil, fat, petroleum ether and most hydrocarbons.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
MDL Number MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Synonym triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanolShow MoreShow Less
PubChem CID 8172
ChEBI CHEBI:44926
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethanol
SMILES OCCOCCOCCO

Specifications

Melting Point -7°C
Density 1.125
Boiling Point 284°C to 288°C
Flash Point 165°C (329°F)
Odor Nearly odorless
Linear Formula H(OCH2CH2)3OH
Refractive Index 1.4555
Quantity 2500 mL
Beilstein 969357
Sensitivity Hygroscopic
Merck Index 14,9670
Solubility Information Miscible with water,ethanol,acetone,benzene,toluene,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil,fat,petroleum ether and most hydrocarbons.
Formula Weight 150.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethylene glycol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250755 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA10772.AP,A10772.0D,A10772.0F
Lot Number10245697 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA10772.AP,A10772.0D,A10772.0F
Lot NumberT24J075 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog NumberA10772.AP,A10772.0D,A10772.0F
Lot Number10245501 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA10772.AP,A10772.0D,A10772.0F
Lot NumberM12J033 Certificate TypeCertificate of Analysis Date23/01/2023 Catalog NumberA10772.AP,A10772.0D,A10772.0F

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethylene glycol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.