missing translation for 'onlineSavingsMsg'
Learn More
Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals Product Code.: 11437057

Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals

Product Code. 11437057
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11437057

Brand: Thermo Scientific Alfa Aesar A14420.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of μ -amino esters from lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.

Solubility
Soluble in dichloromethane.

Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxoniumShow MoreShow Less
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Melting Point 84°C to 89°C
Assay Percent Range 95%, Stabilized with 3-5% diethyl ether
Linear Formula (CH3CH2)3OBF4
Quantity 500 g
UN Number UN2925
Beilstein 3598090
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane.
Formula Weight 189.99
Percent Purity 95%
Chemical Name or Material Triethyloxonium tetrafluoroborate, stabilized with 3 to 5% diethyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252758 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberA14420.22,A14420.14,A14420.36,A1442002
Lot Number10249620 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog NumberA14420.22,A14420.14,A14420.36,A1442002
Lot Number10249594 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog NumberA14420.22,A14420.14,A14420.36,A1442002
Lot Number10248996 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog NumberA14420.22,A14420.14,A14420.36,A1442002
Lot Number10246795 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog NumberA14420.22,A14420.14,A14420.36,A1442002

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.