Learn More
Trimethoxysilane, 95%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 2487-90-3 |
---|---|
Molecular Formula | C3H10O3Si |
Molecular Weight (g/mol) | 122.2 |
MDL Number | MFCD00008340 |
InChI Key | PZJJKWKADRNWSW-UHFFFAOYSA-N |
Synonym | trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook reference |
PubChem CID | 6327428 |
IUPAC Name | trimethoxysilicon |
SMILES | CO[Si](OC)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10106613
|
Thermo Scientific Acros
392170050 |
5 g | Glass bottle |
N/A
|
N/A | ||||
10540164
|
Thermo Scientific Acros
392170250 |
25 g | Glass bottle |
N/A
|
N/A | ||||
10014293
|
Thermo Scientific Acros
392171000 |
100 g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
2487-90-3 | |
122.2 | |
PZJJKWKADRNWSW-UHFFFAOYSA-N | |
6327428 | |
CO[Si](OC)OC |
C3H10O3Si | |
MFCD00008340 | |
trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook reference | |
trimethoxysilicon |
Specifications
2487-90-3 | |
Colorless | |
86°C | |
Authentic | |
C3H10O3Si | |
1.3570 to 1.3600 (20°C, 589nm) | |
0.96 | |
Solubility in water: reacts | |
CO[Si](OC)OC | |
122.2 | |
122.2 | |
Liquid |
-115°C | |
0.9600g/mL | |
-9°C | |
94% min. (GC) | |
HSi(OCH3)3 | |
MFCD00008340 | |
trimethoxysilane, silane, trimethoxy, trimethoxy silane, unii-v1j39xpf91, c3h10o3si, trimethoxysilyl, silanem trimethoxy, trimethoxysilyl radical, 4-01-00-01266 beilstein handbook reference | |
PZJJKWKADRNWSW-UHFFFAOYSA-N | |
trimethoxysilicon | |
6327428 | |
95% | |
Trimethoxysilane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Fatal if inhaled.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-637-2
- Trimethoxysilane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H261-In contact with water releases flammable gases.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only