missing translation for 'onlineSavingsMsg'
Learn More
Trimethyl borate, 99%, Thermo Scientific Chemicals Product Code.: 10061500

Trimethyl borate, 99%, Thermo Scientific Chemicals

Product Code. 10061500
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10061500

Brand: Thermo Scientific Acros 140121000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-43-7
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3Show MoreShow Less
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34.0°C
Color Colorless
Density 0.9150g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C3H9O3B
Linear Formula (CH3O)3B
Refractive Index 1.3562 to 1.3582
Quantity 100 g
Beilstein 01, 287
Fieser 01,673; 13,218
Merck Index 15, 9882
Specific Gravity 0.915
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Molecular Weight (g/mol) 103.91
Viscosity 0.36 mPa.s (25°C)
Formula Weight 103.91
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethyl borate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449797 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number140125000,140120025,140121000
Lot NumberA0447000 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number140125000,140120025,140121000
Lot NumberA0410916 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number140125000,140120025,140121000
Lot NumberA0417461 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number140125000,140120025,140121000
Lot NumberA0445998 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog Number140125000,140120025,140121000
compliance-icons compliance-icons
Product Identifier
  • Trimethyl borate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trimethyl borate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.