Learn More
Trimethyl borate, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | -34.0°C |
Color | Colorless |
Density | 0.9150g/mL |
Boiling Point | 68.0°C to 69.0°C |
Flash Point | −8°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C3H9BO3 |
Linear Formula | (CH3O)3B |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0449797 | Certificate of Analysis | 31/07/2024 | 140125000,140120025,140121000 |
A0447000 | Certificate of Analysis | 29/04/2024 | 140125000,140120025,140121000 |
A0410916 | Certificate of Analysis | 13/12/2022 | 140125000,140120025,140121000 |
A0417461 | Certificate of Analysis | 12/12/2022 | 140125000,140120025,140121000 |
A0445998 | Certificate of Analysis | 23/09/2022 | 140125000,140120025,140121000 |
Safety and Handling


- Trimethyl borate
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Trimethyl borate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.