Learn More
Tris Base (White Crystals or Crystalline Powder/Triple-Crystallized), Fisher BioReagents™
For sensitive molecular biology, tissue culture, and electrophoresis methods
Brand: Fisher BioReagents BP154-1
163.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
For sensitive molecular biology, tissue culture, and electrophoresis methods
This ultrapure, triple-crystallized buffer component is alcohol-free, so it is compatible with sensitive molecular biology, tissue culture, and electrophoresis methods.
Specifications
0.05 max. (1M solution) at 280nm | |
Alcohol-Free | |
168.5°C | |
White | |
Powder or Solid | |
DNase free | |
≥99.9 % | |
C4H11NO3 | |
Poly Bottle | |
15, 9951 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 |
Tris Base | |
77-86-1 | |
219°C | |
10.4 | |
1 kg | |
DNase-, RNase- and Protease-Free | |
Pass Test | |
H2NC(CH2OH)3 | |
Protease free | |
Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
≥99.9% |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Tris(hydroxymethyl)aminomethane
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
22Minutes
29Seconds
59 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
22Minutes
29Seconds
59
Your input is important to us. Please complete this form to provide feedback related to the content on this product.