missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3 Product Code.: 10611662

Thermo Scientific Chemicals Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3

Product Code. 10611662
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 10611662

Brand: Thermo Scientific Chemicals 327330010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610769-35-2 | C14H30BN3O14 | 475.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent, pH buffer
TRUSTED_SUSTAINABILITY

Specifications

CAS 610769-35-2
Color Colorless
pH 8.2 to 8.4 (25°C)
Physical Form Liquid
Quantity 1 L
Molecular Formula C14H30BN3O14
MDL Number MFCD00236358
Packaging Plastic bottle
Beilstein 04, 303
Merck Index 14, 9772
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane
InChI Key OSBLTNPMIGYQGY-UHFFFAOYSA-N
SMILES B(O)(O)O.C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.C(C(CO)(CO)N)O
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;boric acid
Molecular Weight (g/mol) 475.211
PubChem CID 21257725
Grade Molecular Biology
Chemical Name or Material Tris Borate EDTA buffer, pH 8.3
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459522 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number327330010
Lot NumberA0459522 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number327330010
Lot NumberA0389927 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number327330010
Lot NumberA0404611 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number327330010
Lot NumberA0441452 Certificate TypeCertificate of Analysis Date29/09/2022 Catalog Number
compliance-icons
Product Identifier
  • Tris Borate EDTA buffer, 10x, DNase, RNase and Protease free
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Boric acid, EDTA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Tris Borate EDTA buffer, for molecular biology, 10x, DNAse, RNAse and Protease free, pH 8.3 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.