Learn More
Interscience™ Trocar for Broth Bags
Trocar compatible with most tubings.
Brand: Interscience™ 513020
This item is not returnable.
View return policy
Description
- Can be used with all tubings except those with an inner diameter of 1.6 mm
Specifications
Trocar | |
Autoclavable | |
Drawing Fluid from Bag | |
DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
Interscience™ Trocar for Broth Bags > Trocar for Broth Bags; For Use With (Equipment): DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro
Spot an opportunity for improvement?Share a Content Correction