missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptone Soya Broth (Bottled)

Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.

Brand:  Thermo Scientific™ BO0369M

51.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12693428

  • £102.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Grow a wide variety of microorganisms with Thermo Scientific™ Oxoid™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.

  • Conforms to formulations detailed in International Pharmacopoeia.
  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Straw, Clear
Tryptone Soya Broth (Bottled)
United Kingdom
100.0g + 1.5g
10/Pk.
2 to 25°C
Vial Bottle
Agar
10 vial bottles with narrow septum, injectable closures in a box.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4500589 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberBO0369M
Lot Number4499360 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberBO0369M
Lot Number4497720 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberBO0369M
Lot Number4494486 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberBO0369M
Lot Number4489670 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberBO0369M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Tryptone Soya Broth (Bottled) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.