Learn More
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core are used to retrieve stir bars from mixing containers. The PTFE stir bar retriever is chemically resistant, inert, and leak-free. Uses Samarium-Cobalt magnets at its core.
Brand:  Fisherbrand™ 004.2450.RE
 View more versions of this product
Product Code. 16245871
  • £50.90 / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inert and leak-free
  • Molded in pure, FDA-approved PTFE
  • Alnico V magnets
  • Autoclavable
  • For girant stirrer bars
Specifications
Specifications
White
For the removal of Giant Stir Bars from vessels of all kinds
450 mm
1
Turbo PTFE Stir Bar Retriever with Rare Earth Core for Giant Stir Bars
18 mm
PTFE with Rare Earth Magnet Core
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Turbo PTFE Stir Bar Retriever with Rare Earth Core > Length: 450mm; Color: White; Material: PTFE; Core: RareEarth Magnet