Learn More
Thermo Scientific Chemicals Tween 20 washing buffer, 1% in PBS (10X)
Brand: Thermo Scientific Chemicals J63314.K2
67.12 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Buffer | |
Viscous Liquid | |
Keep cold |
1 L | |
1% in PBS (10X) |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y18K556 | Certificate of Analysis | 26/11/2024 | J63314.K3,J63314.K2,J63314.AP |
X01K553 | Certificate of Analysis | 10/10/2024 | J63314.K3,J63314.K2,J63314.AP |
W16K502 | Certificate of Analysis | 27/09/2024 | J63314.K3,J63314.K2,J63314.AP |
W16K501 | Certificate of Analysis | 25/09/2024 | J63314.K3,J63314.K2,J63314.AP |
U06K506 | Certificate of Analysis | 16/08/2024 | J63314.K3,J63314.K2,J63314.AP |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
16Hours
15Minutes
26Seconds
47
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only