Learn More
DuPont™ Tychem™ 2000C Gown
312.41 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Medium
2X-Large
Description
- Double cuffs for increased protection and tighter connection with gloves
- Elasticated wrists
- Knitted cuff (inside) for better comfort
- Offers good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure)
- Used for splash or pressurised splash protection in a variety of industrial environments, including pulp and paper manufacturing, food processing, chemical processing and pharmaceutical manufacturing
- Disposable
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | Medium |
Color | Yellow |
Disposable | Single-use |
Certifications/Compliance | (EU) 2016/425, EN 14126, EN 1149-1 |