missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ Lab Coats, Zipper
Offers enhanced protection for body parts that are more exposed to hazardous substances. Lab coats are composed of flash spun high density polyethylene, providing an idea balance of protection, durability and comfort.
£70.50 - £336.96
Specifications
Product Type | Chemical Resistant Lab Coat |
---|---|
Gender | Unisex |
Disposable | Limited-use |
Sterility | Non-sterile |
Resistance | Chemicals |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11925864
|
DuPont™
D13398951 |
Large |
£331.00
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||||
12332249
|
DuPont™
D13495983 |
£70.50
Pack of 10 |
In Stock
Log in to see stock. |
|||||||
11723123
|
DuPont™
D13496004 |
Small |
£336.96
Pack of 50 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
12362249
|
DuPont™
D13495948 |
2X-Large |
£73.32
Pack of 10 |
In Stock
Log in to see stock. |
||||||
12352249
|
DuPont™
D13398984 |
X-Large |
£72.26
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Silicon non-added
- With collar, zipper closure and 2 pockets
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Chemical Resistant Lab Coat | |
Limited-use | |
Chemicals |
Unisex | |
Non-sterile | |
Pharmaceutical, Lead and Asbestos Abatement/Remediation, General Maintenancce/Operations, Spray Painting, Clean-up |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuPont™ Tyvek™ Lab Coats, Zipper