Learn More
Invitrogen™ UltraComp eBeads™ Compensation Beads
Designed for use in compensation with all fluorochromes excited by ultraviolet, violet, blue, green, yellow-green, and red lasers. UltraComp eBeads™ react with antibodies of mouse, rat, and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test.
£96.75 - £289.00
Specifications
Product Line | eBeads™ |
---|---|
Product Type | Compensation Beads |
For Use With (Equipment) | Flow Cytometer |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
15385356
|
Invitrogen™
01-2222-42 |
100 tests |
£289.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
16881320
|
Invitrogen™
01-2222-42 |
100 tests | N/A | N/A | N/A | |||||
15536296
|
Invitrogen™
01-2222-41 |
25 tests |
£96.75
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
- This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
- Compatible with all fluorochromes excited by an ultraviolet (355nm) or violet (405nm) laser
- Should be used with standard staining buffers which contain PBS or HBSS, protein such as BSA or FBS, and sodium azide. No other additives should be used.
Specifications
eBeads™ | |
Compensation Beads | |
Flow Cytometer |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.