missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals UNC2025, 95% Product Code.: 17140636

Thermo Scientific Chemicals UNC2025, 95%

Product Code. 17140636
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 17140636

Brand: Thermo Scientific Chemicals 467881000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1429881-91-3 | C28H40N6O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1429881-91-3
Assay Percent Range 95%
Quantity 100 mg
Molecular Formula C28H40N6O
Formula Weight 476.66
Chemical Name or Material UNC2025

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number344433 Certificate TypeCertificate of Analysis Date28/09/2023 Catalog Number467881000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals UNC2025, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.