Learn More
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD
Anthracite trainers made of black water repellent and breathable knitted fabric, slip resistant PU/TPU outsole, steel toe cap
Brand: ABEBA™ 31775/46
This item is not returnable.
View return policy
Description
- Made for comfort and safety
- Comply to EN ISO 20345: 2012, S2, SRC
- ESD dissipative according EN 61340
- Slip resistant PU/TPU outsole
- Breathable and water repellent knitted fabric
- Steel toe cap (200 J)
- Breathable and antibacterial lining with silver ions
- Removable insole active comfort
Specifications
Anthracite | |
46 | |
ESD according 61340 class climatic 1, EN ISO 20345: 2012 S2 SRC | |
Trainers | |
S2 SRC ESD |
Knitted Fabric | |
Steel | |
Unisex | |
Water Repellent |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD > Size: 46
Spot an opportunity for improvement?Share a Content Correction