missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Venlafaxine hydrochloride
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 1 g |
Solubility | Solubility aq. soln.: slightly soluble. |
Melting Point | 217.0°C |
Formula Weight | 313.86 |
InChI Key | QYRYFNHXARDNFZ-UHFFFAOYNA-N |
SMILES | [H+].[Cl-].COC1=CC=C(C=C1)C(CN(C)C)C1(O)CCCCC1 |
IUPAC Name | hydrogen 1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol chloride |
Molecular Weight (g/mol) | 313.87 |
Chemical Name or Material | Venlafaxine hydrochloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling

Product Identifier
- Venlafaxine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Venlafaxine hydrochloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Venlafaxine hydrochloride >
Spot an opportunity for improvement?Share a Content Correction