missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Vermiculite Packing Granules

Contains hazardous liquid spills in a sand-like form. Azpack™ Vermiculite Packing Granules can also be used as garden fill, loft insulation or mix with cement to form lightweight concrete.

Brand:  Azpack™ 12387238

Product Code. 12387238

  • £143.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ultra lightweight material for packing hazardous chemicals and absorbing spills
  • Non-combustible granules are insoluble in water and all organic solvents
  • Absorbs and holds a wide range of liquids
  • Granule size ranges nominally between 3 to 15mm
  • Supplied in 4 cubic foot bags (approx 100 litres)
  • Bag weighs 8 kilos.
Specifications

Specifications

100 L
Absorbent Granules
1 Ea.
8 kg
Ultra lightweight granules
Bag
3 to 15mm (granules)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azpack™ Vermiculite Packing Granules > Size: 3 to 15 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.