missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
£35.05 - £301.11
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15685350
|
Honeywell-Chromasolv
39253-250ML |
250 mL | N/A | ||||||
15605360
|
Honeywell-Chromasolv
39253-500ML |
500 mL | N/A | ||||||
15665350
|
Honeywell-Chromasolv
39253-1L |
1 L |
£59.57
1 L |
In Stock
Log in to see stock. |
|||||
16225301
|
Honeywell-Riedel-de Haen
39253-2.5L |
2.5 L | N/A | ||||||
16419019
|
Honeywell-Riedel-de Haen
39253-2.5L |
2.5 L |
£66.46
Each |
In Stock
Log in to see stock. |
|||||
15608400
|
Honeywell-Chromasolv
39253-4L |
4 L |
£80.28
4 L |
In Stock
Log in to see stock. |
|||||
15695350
|
Honeywell-Chromasolv
39253-4X4L |
4 x 4 L |
£301.11
Pack of 4 |
In Stock
Log in to see stock. |
|||||
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
max. 0.022 at 210 nm, max. 0.004 at 230 nm | |
100°C | |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
LC-MS | |
1.000 g/mL at 3.98 °C(lit.) |
7732-18-5 | |
100.0000% | |
H2O | |
2050024 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18 | |
Glass Bottle | |
Water, CHROMASOLV™ LC-MS |
Safety and Handling
missing translation for 'shelfLife' : 547 days from date of manufacture
missing translation for 'einecsNumber' : 231-791-2