missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WEB 2086, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
Description
Potent, selective platelet-activating factor (PAF) receptor antagonist (Ki = 16.3 nM). Displays anti-inflammatory, antiangiogenic and anticancer activity. Inhibits growth and proliferation of MCF-7 breast cancer cells.
Specifications
Quantity | 1 mg |
Formula Weight | 455.96 |
Percent Purity | >99% |
Chemical Name or Material | WEB 2086 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'