Learn More
Thermo Scientific™ Webseal 384-Well Plate
384-Well Plate with square v-shaped base.
Brand:  Thermo Scientific™ 60180-P109B
62.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17483193
  • £68.83 / Pack of 10
Explore more special offers
Description
Description
  • 384-well
  • Polypropylene
  • Square V Base
  • Barcoded
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Without Lid
Webseal
Single-use
Polypropylene
384
Conical Bottom (V-well)
Autoclavable
Clear
Full
145 μL
Non-sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Webseal 384-Well Plate > 145 μL