Learn More
Welch™ DuoSeal™ Pump Oil
For use in DuoSeal High-Vacuum Pumps
Brand:  Welch™ U1407K-15
 View more versions of this product
Product Code. 12835353
  • £104.00 / 3.70 L
Explore more special offers
Description
Description
For use in DuoSeal High-Vacuum Pumps
For use in DuoSeal High-Vacuum Pumps. One gallon.Specially fractionate oil has properties enabling highest performance for DuoSeal pumps
  • Low vapor Pressure
  • High Viscosity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vaccum Pump Oil
DuoSeal™ Belt-driven pumps
1 gal.
Provides the lubrication that helps enable renowned DuoSeal™ pump durability and ensure the highest vacuum performance
1 Ea.
3.78 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch™ DuoSeal™ Pump Oil > 1 gal. (4L)