Learn More
Cytiva Whatman™ PTFE Membrane Circles
Provides clarification of corrosives, solvents, and aggressive fluids. Whatman™ PTFE Membrane Circles are particularly useful in HPLC analysis when solid particles can cause permanent damage to the column.
£223.00 - £1020.00
Product Code | Brand | Diameter (Metric) | Pore Size | Membrane Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Membrane Type | Price | Quantity & Availability | |||
11383435
|
Cytiva
10411311 |
47 mm | 0.45 μm | TE 36 |
£414.00
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
11363445
|
Cytiva
10411113 |
50 mm | 5 μm | TE 38 |
£286.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11353445
|
Cytiva
10411111 |
47 mm | 5 μm | TE 38 |
£283.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11373445
|
Cytiva
10411116 |
90 mm | 5 μm | TE 38 |
£487.00
Pack of 25 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
11343445
|
Cytiva
10411108 |
37 mm | 5 μm | TE 38 |
£277.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11333445
|
Cytiva
10411130 |
150 mm | 5 μm | TE 38 |
£1020.00
Pack of 25 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11313445
|
Cytiva
10411211 |
47 mm | 1.0 μm | TE 37 |
£280.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11303445
|
Cytiva
10411205 |
25 mm | 1.0 μm | TE 37 |
£223.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11393435
|
Cytiva
10411313 |
50 mm | 0.45 μm | TE 36 |
£286.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11345404
|
Cytiva
10411413 |
50 mm | 0.2 μm | TE 35 |
£313.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11335404
|
Cytiva
10411411 |
47 mm | 0.2 μm | TE 35 |
£307.00
Pack of 50 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11323445
|
Cytiva
10411213 |
50 mm | 1.0 μm | TE 37 |
£286.00
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11325404
|
Cytiva
10411405 |
25 mm | 0.2 μm | TE 35 |
£244.00
Pack of 50 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
11373435
|
Cytiva
10411305 |
25 mm | 0.45 μm | TE 36 |
£223.00
Pack of 50 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||
Description
Provides clarification of corrosives, solvents, and aggressive fluids. Whatman™ PTFE Membrane Circles are particularly useful in HPLC analysis when solid particles can cause permanent damage to the column.
- Hydrophobic
- Chemically stable and inert
- Can be used up to 120°C
- Laminated onto randomly arranged polypropylene support material
- For filtration with organic and chemically aggressive solutions or gases
- Particularly suitable for HPLC analysis
Specifications
PTFE | |
Hydrophobic |