Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Seals
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand: DWK Life Sciences W988010-S
710.59 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Sterile
- Flip-off Seal
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications
Blue | |
Sterile | |
Wheaton CompletePAK | |
USP 788, 85, and 71, Container Closure Integrity (CCI) certified | |
2 mL Serum Vial |
Aluminum | |
285/Cs. | |
Sterile Flip Seal | |
13 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.