Learn More
DWK Life Sciences Wheaton™ I-LOC Closures
Eliminate use of separate aluminum seal and rubber stopper
Brand:  DWK Life Sciences 240676-04
52.86 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12408732
  • £71.60 / Pack of 100
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminate use of separate aluminum seal and rubber stopper
  • Polypropylene closures
  • Gray bromobutyl stoppers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bromobutyl
Polypropylene
Screw Cap
Blue
Autoclavable
20-400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ I-LOC Closures > Blue