Learn More
DWK Life Sciences Wheaton™ I-LOC Closures
Eliminate use of separate aluminum seal and rubber stopper
Brand:  DWK Life Sciences 240676-05
54.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12418732
  • £73.55 / Pack of 100
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminate use of separate aluminum seal and rubber stopper
  • Polypropylene closures
  • Gray bromobutyl stoppers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bromobutyl
Polypropylene
Screw Cap
Yellow
Autoclavable
20-400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ I-LOC Closures > Yellow