Learn More
Corning™ XT Freezing Core for CoolBox™ XT Workstation

Allows for continuous uninterrupted cooling. Corning™ XT Freezing Core for CoolBox™ XT Workstation provides cooling source in CoolBox XT Workstation when thermo-conductive CoolRack or CoolSink tube and plate module is placed on top.
Brand: Corning™ 432082
237.09 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
XT Freezing Core for CoolBox XT, for below 0°C, a second one allows for continuous, uninterrupted cooling.

Specifications
XT Freezing Core | |
1/Pk. | |
Maintaining sample temperature | |
16.2 x 12.1 x 6.1 cm (6.3 x 4.7 x 2.1 in.) |
Blue | |
1/Cs. | |
CoolBox XT Workstation |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.