Learn More
Cytiva HyClone™ AdvanceSTEM Cell Culture Media
Provides optimized conditions for embryonic stem cell expansion and differentiation.
Brand: Cytiva SH30822.01
This item is not returnable.
View return policy
Description
Specifications
AdvanceSTEM IMDM4SC without L-glutamine | |
Liquid | |
No Glutamine |
Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) | |
Sodium Pyruvate | |
500 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Cytiva HyClone™ AdvanceSTEM Cell Culture Media > Description: AdvanceSTEM IMDM4SC without L-glutamine, dimQuantity: 500 mL
Spot an opportunity for improvement?Share a Content Correction