missing translation for 'onlineSavingsMsg'
Learn More
Xylene, Technical, Fisher Chemical™ Product Code.: 10691373

Xylene, Technical, Fisher Chemical™

Product Code. 10691373
2.5 L, Plastic-coated Glass Bottle
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10691373

Brand: Fisher Chemical X/0100/PB17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number 77264
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -34°C
Color Colorless
Boiling Point 136°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Formula Weight 106.17g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2526020 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberX/0100/PB17,X/0100/25,X/0100/27,X/0100/17
Lot Number2526026 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberX/0100/PB17,X/0100/25,X/0100/27,X/0100/17
Lot Number2526025 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberX/0100/PB17,X/0100/25,X/0100/27,X/0100/17
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aromatic hydrocarbons, C8; Light Oil Redistillate, high boiling, Xylenes (o-, m-, p- isomers), Ethyl
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylene, Technical, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.