missing translation for 'onlineSavingsMsg'
Learn More
Xylenes, 98%, pure, mixture of isomers, Thermo Scientific Chemicals Product Code.: 10370394

Xylenes, 98%, pure, mixture of isomers, Thermo Scientific Chemicals

Product Code. 10370394
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10370394

Brand: Thermo Scientific Acros 383930010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7, 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzene

Specifications

Melting Point -34.0°C
Density 0.8650g/mL
Boiling Point 136.0°C to 140.0°C
Quantity 1 L
Flash Point 23°C
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 98%
Residue after Evaporation 0.01% max.
Specific Gravity 0.865
Linear Formula C6H4(CH3)2
Viscosity 0.6 mPa.s (20°C)
Water 0.05% max. (K.F.)
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity 98%
Grade Pure
Chemical Name or Material Xylenes, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2411796 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10370394,383930050,383930010,383930250
Lot Number2411798 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10370394,383930050,383930010,383930250
Lot Number2409248 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10370394,383930050,383930010,383930250
Lot Number2411794 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10370394,383930050,383930010,383930250
Lot Number2411793 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10370394,383930050,383930010,383930250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylenes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xylenes, 98%, pure, mixture of isomers, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.