missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Y-PER™ Plus Dialyzable Yeast Protein Extraction Reagent

Extract and solubilize proteins for analysis from yeast and bacterial cells with this effective, fully dialyzable lysis reagent.

Brand:  Thermo Scientific™ 78999

Product Code. 10197773

  • £360.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Effectively lyse yeast and bacterial cells to extract and solubilize proteins for analysis with Thermo Scientific™ Y-PER-Plus Dialyzable Yeast Protein Extraction Reagent.

The proprietary formulation of cell lysis buffer facilitates protein extraction from S. cerevisiae and S. pastoris yeast cell cultures. Glass bed disruption is not necessary; protein extraction is achieved by a 20-minute incubation in the reagent at room temperature. Unlike other cell lysis buffers, the Y-PER-Plus Reagent uses a detergent component that is fully dialyzable, allowing complete exchange of the extracted protein into any buffer required by downstream techniques.

Highlights:

  • Stable formulation designed for room temperature storage
  • Yields protein extract with simple 20-minute hands-off incubation
  • Requires no rigorous mechanical disruption or lengthy enzymatic treatments
  • Tris-based buffer has a very low ionic strength and does not include sodium chloride
  • Simple formulation contains no reducing agents or chelating agents
  • Fully dialyzable formulation provides complete control over downstream applications
  • Compatible with both the Thermo Scientific Pierce™ BCA and Coomassie Plus™ Protein Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL
Yeast Protein Extraction Reagent
Upon receipt store at room temperature. Product shipped at ambient temperature.
Liquid
500 mL
Y-PER™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ382453 Certificate TypeCertificate of Analysis Date26/10/2024 Catalog Number78999
Lot NumberYJ374909 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number78999
Lot NumberYD366255 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog Number78999
Lot NumberYB359229 Certificate TypeCertificate of Analysis Date28/02/2023 Catalog Number78999
Lot NumberXI352171 Certificate TypeCertificate of Analysis Date30/09/2022 Catalog Number78999
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Y-PER™ Plus Dialyzable Yeast Protein Extraction Reagent > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.