missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antiepileptic that possesses a broad spectrum anticonvulsant and mechanistic profile. Blocks voltage-sensitive Na+ and T-type Ca2+ channels, stimulates BKCa channels, modulates GABA, glutamate and monoamine neurotransmission, inhibits lipid peroxidation and scavenges hydroxyl and nitric oxide free radicals. Displays neuroprotective and antiParkinsonian activity.

Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 212.22 |
Percent Purity | >99% |
Chemical Name or Material | Zonisamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zonisamide, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction