missing translation for 'onlineSavingsMsg'
Learn More
1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals Product Code.: 11351277

1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals

Product Code. 11351277
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11351277

Brand: Thermo Scientific Alfa Aesar L01421.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22013-33-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00006824
InChI Key BZKOZYWGZKRTIB-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-6-amine, 3,4-ethylenedioxyaniline, 6-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-6-amine, 6-amino-1,4-benzodioxan, 1,4-benzodioxan-6-ylamine, 1,4-benzodioxin-6-amine, 2,3-dihydro, 3,4-ethylenedioxy aniline, 2,3-dihydro-benzo 1,4 dioxin-6-ylamine, 2,3-dihydro-1,4-benzodioxin-6-aminShow MoreShow Less
PubChem CID 89148
IUPAC Name 2,3-dihydro-1,4-benzodioxin-6-amine
SMILES NC1=CC=C2OCCOC2=C1

Specifications

Melting Point 26°C to 30°C
Density 1.231
Boiling Point 116°C to 118°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.599
Quantity 100 g
UN Number UN2811
Beilstein 6447
Sensitivity Air and light sensitive
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 1,4-Benzodioxan-6-amine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238328 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberL01421.06,L01421.14,L01421.22
Lot NumberF05M30 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01421.06,L01421.14,L01421.22
Lot NumberB05N20 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01421.06,L01421.14,L01421.22
Lot Number10195336 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01421.06,L01421.14,L01421.22
Lot Number10141032 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01421.06,L01421.14,L01421.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Benzodioxan-6-amine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.