missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthoyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11411778

1-Naphthoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11411778
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11411778

Brand: Thermo Scientific Alfa Aesar A14460.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, amines, alcohols, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879-18-5
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.63
MDL Number MFCD00004002
InChI Key NSNPSJGHTQIXDO-UHFFFAOYSA-N
Synonym 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chlorideShow MoreShow Less
PubChem CID 70146
IUPAC Name naphthalene-1-carbonyl chloride
SMILES ClC(=O)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 16°C to 19°C
Density 1.265
Boiling Point 118°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6525
Quantity 50 g
UN Number UN3265
Beilstein 775785
Sensitivity Moisture Sensitive
Solubility Information Reacts with water.
Formula Weight 190.63
Percent Purity 98%
Chemical Name or Material 1-Naphthoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0447081 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14460.18,A14460.09,A14460.30
Lot NumberA0446849 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberA14460.18,A14460.09,A14460.30
Lot NumberA0433454 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog NumberA14460.18,A14460.09,A14460.30
Lot Number10153088 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14460.18,A14460.09,A14460.30
Lot Number10180709 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14460.18,A14460.09,A14460.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthoyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.