missing translation for 'onlineSavingsMsg'
Learn More
Ebselen, Thermo Scientific™ Product Code.: 15465449

Ebselen, Thermo Scientific™

Product Code. 15465449
10mg
Click to view available options
Quantity:
10mg
This item is not returnable. View return policy

Product Code. 15465449

Brand: Thermo Scientific Alfa Aesar J63190.MA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ebselen inhibits 5-LO, CO, NADPH oxidize, and protein kinase C. It is a potent antioxidant. It possesses a unique glutathione peroxidase-like activity. It is also shown to be an inhibitor of free radical induced apoptosis and neuroprotective agent.

Solubility
Soluble in ethanol, methanol, acetone, acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 60940-34-3
Molecular Formula C13H9NOSe
MDL Number MFCD00210937
Quantity 10mg
UN Number UN3283
Merck Index 14,3486
Synonym ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
Solubility Information Soluble in ethanol,methanol,acetone,acetonitrile.
InChI Key DYEFUKCXAQOFHX-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2
IUPAC Name 2-phenyl-1,2-benzoselenazol-3-one
Molecular Weight (g/mol) 274.192
PubChem CID 3194
ChEBI CHEBI:77543
Formula Weight 274.2
Chemical Name or Material Ebselen
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP11L052 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog NumberJ63190.MA
Lot NumberM29L023 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberJ63190.MA
Lot NumberZ24K036 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ63190.#0,J63190.MA
Lot NumberN14J013 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberJ63190.#0,J63190.MA
Lot NumberT07J025 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberJ63190.#0,J63190.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ebselen, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.