missing translation for 'onlineSavingsMsg'
Learn More
Ebselen, 99% Product Code.: 10512281

Ebselen, 99%

Product Code. 10512281
250 mg, Glass bottle
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10512281

Brand: Thermo Scientific Acros 300572500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60940-34-3 | C13H9NOSe | 274.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 60940-34-3
Melting Point 179°C to 184°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C13H9NOSe
MDL Number MFCD00210937
Quantity 250 mg
Beilstein 27, II, 222
Merck Index 15, 3533
Synonym ebselen, 2-phenyl-1,2-benzisoselenazol-3 2h-one, ebselene, ebseleno, ebselenum, ebselene french, ebselenum latin, ebselen inn, ebseleno spanish, harmokisane
InChI Key DYEFUKCXAQOFHX-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N2C(=O)C3=CC=CC=C3[Se]2
IUPAC Name 2-phenyl-1,2-benzoselenazol-3-one
Molecular Weight (g/mol) 274.192
PubChem CID 3194
ChEBI CHEBI:77543
Formula Weight 274.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Ebselen, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0265701 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number300572500
Lot NumberA0265701 Certificate TypeCertificate of Analysis Date22/07/2022 Catalog Number300572500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ebselen, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.