missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A Tyvek™ Cylindrical Equipment Cover
GREENER_CHOICE

1422A Tyvek™ Cylindrical Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.

£994.00 - £4565.00

Specifications

Product Type Cover
Autoclavable Autoclavable
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
View More Specs

Products 17
Product Code Brand Diameter (English) Height (English) Closure Type Price Quantity & Availability  
Product Code Brand Diameter (English) Height (English) Closure Type Price Quantity & Availability  
17758267
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-1460
14 in. 60 in. None
£2680.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17768267
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-1460E
14 in. 60 in. Elastic
£2745.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17788267
GREENER_CHOICE
View Documents Promotion Details
Keystone Cleanroom Products
COV-TVK-2048
20 in. 48 in. None
£2560.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17798267
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-2048E
20 in. 48 in. Elastic
£2625.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17796889
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-0905E
23 in. 5 in. Heat Seal
£994.00
Pack of 100
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
 
17871721
View Documents
Keystone Cleanroom Products
2620TWELNT
26 in. 20 in. Elastic
£1990.00
Pack of 50
Estimated Shipment: 29-05-2025
to see stock.
Add to basket
 
17778277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-3230
32 in. 30 in. None
£2680.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17788277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-3230E
32 in. 30 in. Elastic
£2875.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17798277
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-3260
32 in. 60 in. None
£3215.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17778297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-484860E
32 in. 60 in. Elastic
£4170.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17788287
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-4410E
44 in. 10 in. Elastic
£2590.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17748297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-4836
48 in. 36 in. None
£2920.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17758297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-4836E
48 in. 36 in. Elastic
£2975.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17788297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-4884
48 in. 84 in. None
£4365.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17798297
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-4884E
48 in. 84 in. Elastic
£4565.00
Pack of 50
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17738307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-5060
50 in. 60 in. None
£1695.00
Pack of 25
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
17748307
GREENER_CHOICE
View Documents
Keystone Cleanroom Products
COV-TVK-5060E
50 in. 60 in. Elastic
£1790.00
Pack of 25
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
Description

Description

  • Keeps cleaned equipment clean and protected from contamination
  • Breathable
  • Form fitting, fast and consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications

Specifications

Cover
White
Steam Sterilization
Tyvek
Steam
<125°C
Autoclavable
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Keystone Cleanroom Products 1422A Tyvek™ Cylindrical Equipment Cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.