missing translation for 'onlineSavingsMsg'
Learn More
3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals Product Code.: 11423564

3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11423564
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11423564

Brand: Thermo Scientific Alfa Aesar A11229.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.

Solubility
Soluble in water (partly), ethanol, and methanol.

Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2861-28-1
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00014576
InChI Key ODVLMCWNGKLROU-UHFFFAOYSA-N
Synonym 1,3-benzodioxole-5-acetic acid, 3,4-methylenedioxy phenylacetic acid, 2-benzo d 1,3 dioxol-5-yl acetic acid, 3,4-methylenedioxyphenylacetic acid, homopiperonylic acid, benzo-1,3-dioxole-5-acetic acid, 1,3-benzodioxol-5-yl acetic acid, 2-2h-1,3-benzodioxol-5-yl acetic acid, 2-1,3-benzodioxol-5-yl acetic acid, 1,3-benzodioxol-5-ylacetic acidShow MoreShow Less
PubChem CID 76115
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetic acid
SMILES C1OC2=C(O1)C=C(C=C2)CC(=O)O

Specifications

Melting Point 127°C to 130°C
Quantity 25 g
Beilstein 177739
Solubility Information Soluble in water (partly),ethanol,and methanol.
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 3,4- (Methylenedioxy)phenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242831 Certificate TypeCertificate of Analysis Date26/08/2022 Catalog NumberA11229.14,A11229.06,A11229.22
Lot Number10196763 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11229.14,A11229.06,A11229.22
Lot NumberZ28B056 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11229.14,A11229.06,A11229.22
Lot Number10206812 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11229.14,A11229.06,A11229.22
Lot Number10203715 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11229.14,A11229.06,A11229.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-(Methylenedioxy)phenylacetic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.