missing translation for 'onlineSavingsMsg'
Learn More
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals Product Code.: 10346130

3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 10346130
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10346130

Brand: Thermo Scientific 115540050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2478-38-8
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008748
InChI Key OJOBTAOGJIWAGB-UHFFFAOYSA-N
Synonym acetosyringone, 3',5'-dimethoxy-4'-hydroxyacetophenone, 1-4-hydroxy-3,5-dimethoxyphenyl ethanone, 4'-hydroxy-3',5'-dimethoxyacetophenone, 3,5-dimethoxy-4-hydroxyacetophenone, acetosyringon, ethanone, 1-4-hydroxy-3,5-dimethoxyphenyl, 1-4-hydroxy-3,5-dimethoxyphenyl ethan-1-one, acetosyringenin, acetophenone, 4'-hydroxy-3',5'-dimethoxyShow MoreShow Less
PubChem CID 17198
ChEBI CHEBI:2404
IUPAC Name 1-(4-hydroxy-3,5-dimethoxyphenyl)ethanone
SMILES COC1=CC(=CC(OC)=C1O)C(C)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123°C to 128°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H2(OCH3)2COCH3
Quantity 5 g
Beilstein 08,II,444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,acetone,ether
Formula Weight 196.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467149 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number115540250,115540010,115540050
Lot NumberA0459462 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number115540250,115540010,115540050
Lot NumberA0450144 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number115540250,115540010,115540050
Lot NumberA0397683 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number115540250,115540010,115540050
Lot NumberA0393807 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number115540250,115540010,115540050
compliance-icons
Product Identifier
  • 3',5'-Dimethoxy-4'-hydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3',5'-Dimethoxy-4'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.