missing translation for 'onlineSavingsMsg'
Learn More
Anthraquinone, 98%, Thermo Scientific Chemicals Product Code.: 10711734

Anthraquinone, 98%, Thermo Scientific Chemicals

Product Code. 10711734
50 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10711734

Brand: Thermo Scientific Acros 104930500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-65-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.22
MDL Number MFCD00001188
InChI Key RZVHIXYEVGDQDX-UHFFFAOYSA-N
Synonym anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinoneShow MoreShow Less
PubChem CID 6780
ChEBI CHEBI:40448
SMILES O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12

Specifications

Melting Point 283.0°C to 288.0°C
Color Green-Yellow to Tan
Boiling Point 379.0°C to 381.0°C
Flash Point 185°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 50 g
Beilstein 07, 781
Fieser 04,20
Merck Index 15, 679
Solubility Information Solubility in water: insoluble. Other solubilities: soluble 0.44g/100g in alcohol (25°C),2.25 in,boiling alcohol; 0.11g/100g in ether (25°C);,0.61g/100g in chloroform (20°C); 0.26g/100g in,benzene (20°C); 0.30g/100g in toluene (25°
IUPAC Name 9,10-dihydroanthracene-9,10-dione
Formula Weight 208.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Anthraquinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461617 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number104930010,104932500,104930500
Lot NumberA0355824 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number104930010,104932500,104930500
Lot NumberA0452671 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog Number104930010,104932500,104930500
Lot NumberA0414099 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number104930010,104932500,104930500
Lot NumberA0408024 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number104930010,104932500,104930500
compliance-icons compliance-icons
Product Identifier
  • Anthraquinone
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Anthraquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anthraquinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.